Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-6-methylpyrimidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
3435-25-4
C5H5ClN2
128.559
MFCD02322991
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine,4-methyl-6-chloropyrimidine,pyrimidine, 4-chloro-6-methyl,4-chloro-6-methyl-pyrimidine,6-chloro-4-methylpyrimidine,4-chloro-6-methyl-1,3-diazine,pubchem13240,6-methyl-4-chloropyrimidin,ksc269g3h,pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl
3435-25-4
C5H5ClN2
128.559
MFCD02322991
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine,4-methyl-6-chloropyrimidine,pyrimidine, 4-chloro-6-methyl,4-chloro-6-methyl-pyrimidine,6-chloro-4-methylpyrimidine,4-chloro-6-methyl-1,3-diazine,pubchem13240,6-methyl-4-chloropyrimidin,ksc269g3h,pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 250mg
Color Yellow
Beilstein 108656
Formula Weight 128.56
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-methylpyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning