Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-methylpyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number AAH6466703
Click to view available options
Quantity:
250 mg
1 g
5 g
3435-25-4
C5H5ClN2
128.559
MFCD02322991
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl
3435-25-4
C5H5ClN2
128.559
MFCD02322991
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Beilstein 108656
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-6-methylpyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit