Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chloro-6-nitro-m-cresol 98.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 134°C |
Quantity | 25 g |
Formula Weight | 187.58 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 4-Chloro-6-nitro-m-cresol |
Safety and Handling
EINECSNumber : (3)-0826&(3)-0938
TSCA : Yes