Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-6-(trifluoromethyl)pyrimidine, 95%

Click to view available options
Quantity:
250mg
1g
37552-81-1
C5H2ClF3N2
182.53
MFCD04972771
TYSPDLZOMUDHQZ-UHFFFAOYSA-N
4-chloro-6-trifluoromethyl pyrimidine,4-chloro-6-trifluoromethyl-pyrimidine,pyrimidine, 4-chloro-6-trifluoromethyl,pubchem13244,ksc494s0d,6-chloro-4-trifluoromethyl pyrimidine
11542881
4-chloro-6-(trifluoromethyl)pyrimidine
C1=C(N=CN=C1Cl)C(F)(F)F
37552-81-1
C5H2ClF3N2
182.53
MFCD04972771
TYSPDLZOMUDHQZ-UHFFFAOYSA-N
4-chloro-6-trifluoromethyl pyrimidine,4-chloro-6-trifluoromethyl-pyrimidine,pyrimidine, 4-chloro-6-trifluoromethyl,pubchem13244,ksc494s0d,6-chloro-4-trifluoromethyl pyrimidine
11542881
4-chloro-6-(trifluoromethyl)pyrimidine
C1=C(N=CN=C1Cl)C(F)(F)F
Color Colorless to Yellow
Quantity 1g
Formula Weight 182.53
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Chloro-6-(trifluoromethyl)pyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures