Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chloro-L-phenylalanine Hydrochloride 98.0+%, TCI America™
Click to view available options
Quantity:
1 g
5 g
Specifications
Color | White |
Quantity | 1 g |
Formula Weight | 236.09 |
Percent Purity | ≥98.0% (T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 4-Chloro-L-phenylalanine Hydrochloride |
Safety and Handling
TSCA : No