Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-N-methylaniline, 95%

Manufacturer:  Alfa Aesar™ B2396203

Quantity 1g

 View more versions of this product

Catalog No. AAB2396203


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

932-96-7
141.6
UN2810
N-Methyl-2-chloroaniline
C7H8ClN
MFCD00000614
2205846
4-Chloro-N-methylaniline

Specifications

>100°C (212°F)
1g
C7H8ClN
MFCD00000614
2205846
4-Chloro-N-methylaniline
238° to 240°C
932-96-7
141.6
UN2810
N-Methyl-2-chloroaniline
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : CX9857900

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit