Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-methylaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
932-96-7
C7H8ClN
141.598
MFCD00000614
XCEYKKJMLOFDSS-UHFFFAOYSA-N
p-chloro-n-methylaniline,benzenamine, 4-chloro-n-methyl,n-methyl-4-chloroaniline,n-p-chlorobenzyl methylamine,n-methyl-p-chloroaniline,aniline, p-chloro-n-methyl,unii-2ixy9ja2p8,4-chloro-n-methylbenzenamine,ccris 2889,n-4-chlorophenyl-n-methylamine
70272
4-chloro-N-methylaniline
CNC1=CC=C(C=C1)Cl
932-96-7
C7H8ClN
141.598
MFCD00000614
XCEYKKJMLOFDSS-UHFFFAOYSA-N
p-chloro-n-methylaniline,benzenamine, 4-chloro-n-methyl,n-methyl-4-chloroaniline,n-p-chlorobenzyl methylamine,n-methyl-p-chloroaniline,aniline, p-chloro-n-methyl,unii-2ixy9ja2p8,4-chloro-n-methylbenzenamine,ccris 2889,n-4-chlorophenyl-n-methylamine
70272
4-chloro-N-methylaniline
CNC1=CC=C(C=C1)Cl
Refractive Index 1.5830 to 1.5850 (20°C, 589nm)
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula ClC6H4NHCH3
Beilstein 12, 609
Formula Weight 141.6
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 4-Chloro-N-methylaniline, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may darken during storage