Learn More
Accela Chembio IncSupplier Diversity Partner 4-CHLORO-N N-DIMETHYLANIL 1G  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0271001G
4-CHLORO-N,N-DIMETHYLANILINE, 1G, 698-69-1, MFCD07780200, 97+%, RETEST PERIOD: 540DAYS, LIGHT SENSITIVE/NITROGEN OR ARGON/+4 c
Catalog No. 50-649-989