Alfa Aesar™ 4'-Chloroacetophenone, 98+%

Manufacturer: Alfa Aesar™ A121150I

Quantity 5000g

 View more versions of this product

Catalog No. AAA121150I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-91-2
    Molecular Formula C8H7ClO
    Formula Weight 154.6
    MDL Number MFCD00000624
    UN Number UN3416
    Beilstein 386014
    Merck Index 14,2116
    Chemical Name or Material 4'-Chloroacetophenone

    Specifications

    Formula Weight 154.6
    Health Hazard 3 GHS P Statement
    P210-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H7ClO
    Boiling Point 230° to 232°C
    CAS 99-91-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H330-H227-H302-H315-H319-H335
    Fatal if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 90°C (194°F)
    TSCA TSCA
    MDL Number MFCD00000624
    UN Number UN3416
    Beilstein 386014
    Merck Index 14,2116
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID
    RTECS Number KM5600000
    Melting Point 17° to 20°C
    Quantity 5000g
    Chemical Name or Material 4'-Chloroacetophenone
    Assay ≥98%

    Description

    Applications
    Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

    Solubility
    Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.

    GHS H Statement
    H330-H227-H302-H315-H319-H335
    Fatal if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit