Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Chloroacetyl)morpholine, 97+%

Click to view available options
Quantity:
1g
5g
1440-61-5
C6H10ClNO2
163.601
MFCD00721939
YMQRPXBBBOXHNZ-UHFFFAOYSA-N
4-2-chloroacetyl morpholine,morpholine, 4-chloroacetyl,4-chloroacetyl morpholine,n-chloroacetyl morpholine,2-chloro-1-morpholin-4-yl ethan-1-one,n-chloroacetylmorpholine,chloroacetic acid morpholide,2-chloro-1-morpholinoethanone,2-chloro-1-morpholin-4-yl-ethanone,chloroacetic acid, morpholide
74040
2-chloro-1-morpholin-4-ylethanone
C1COCCN1C(=O)CCl
1440-61-5
C6H10ClNO2
163.601
MFCD00721939
YMQRPXBBBOXHNZ-UHFFFAOYSA-N
4-2-chloroacetyl morpholine,morpholine, 4-chloroacetyl,4-chloroacetyl morpholine,n-chloroacetyl morpholine,2-chloro-1-morpholin-4-yl ethan-1-one,n-chloroacetylmorpholine,chloroacetic acid morpholide,2-chloro-1-morpholinoethanone,2-chloro-1-morpholin-4-yl-ethanone,chloroacetic acid, morpholide
74040
2-chloro-1-morpholin-4-ylethanone
C1COCCN1C(=O)CCl
Melting Point 27°C to 30°C
Quantity 5g
UN Number UN3261
Formula Weight 163.61
Percent Purity ≥97%
Chemical Name or Material 4-(Chloroacetyl)morpholine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.