Alfa Aesar™ 4-Chloroanisole, 99%

Manufacturer: Alfa Aesar™ A1137730

Quantity 250g

 View more versions of this product

Catalog No. AAA1137730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 623-12-1
    Molecular Formula C7H7ClO
    Formula Weight 142.59
    MDL Number MFCD00000616
    Beilstein 1858901
    Synonym 4-Methoxychlorobenzene
    Chemical Name or Material 4-Chloroanisole

    Specifications

    Formula Weight 142.59
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Flash Point 78°C (172°F)
    Boiling Point 198° to 200°C
    Molecular Formula C7H7ClO
    Beilstein 1858901
    Synonym 4-Methoxychlorobenzene
    Quantity 250g
    MDL Number MFCD00000616
    CAS 623-12-1
    Health Hazard 1 Warning
    Melting Point −18°C
    Chemical Name or Material 4-Chloroanisole
    Assay 99%

    Description

    Applications
    4-Chloroanisole was found to react with Cu (II)-smectite forming a blue clay-organic complex. 4-Chloroanisole was chosen as the main test substrate for the optimisation studies as it is electronically deactivated and thus resistant to oxidative addition and consequently very reluctant to enter a catalytic manifold.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit