Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzaldehyde, 98.5+%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
104-88-1
C7H5ClO
140.57
AVPYQKSLYISFPO-UHFFFAOYSA-N
p-chlorobenzaldehyde,benzaldehyde, 4-chloro,benzaldehyde, p-chloro,p-chlorobenzenecarboxaldehyde,4-chloro-benzaldehyde,4-chlorobenzenaldehyde,4-chloro benzaldehyde,ccris 857,para-chlorobenzaldehyde,unii-e67727up9z
7726
CHEBI:28105
4-chlorobenzaldehyde
C1=CC(=CC=C1C=O)Cl
104-88-1
C7H5ClO
140.57
AVPYQKSLYISFPO-UHFFFAOYSA-N
p-chlorobenzaldehyde,benzaldehyde, 4-chloro,benzaldehyde, p-chloro,p-chlorobenzenecarboxaldehyde,4-chloro-benzaldehyde,4-chlorobenzenaldehyde,4-chloro benzaldehyde,ccris 857,para-chlorobenzaldehyde,unii-e67727up9z
7726
CHEBI:28105
4-chlorobenzaldehyde
C1=CC(=CC=C1C=O)Cl
Melting Point >46.0°C
Specific Gravity 1.196
Boiling Point 214.0°C
Quantity 5kg
Packaging Plastic drum
Color White to Yellow
Density 1.1960g/mL
Infrared Spectrum Authentic
Flash Point 88°C
Assay Percent Range 98%
Linear Formula ClC6H4CHO
Beilstein 07,235
Solubility Information Solubility in water: 935mg/l (20°C)
Formula Weight 140.57
Physical Form Crystalline Mass
Percent Purity ≥98.5%
Chemical Name or Material 4-Chlorobenzaldehyde
Show More Show Less