Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzamidine hydrochloride, 95%, Acros Organics

Click to view available options
Quantity:
5g
50g
14401-51-5
C7H7ClN2·ClH
191.06
RXAOGVQDNBYURA-UHFFFAOYSA-N
4-chlorobenzamidine hydrochloride,4-chlorobenzene-1-carboximidamide hydrochloride,4-chlorobenzamidine hcl,p-chlorobenzamidine hydrochloride,4-chloro-benzamidine hydrochloride,4-chlorobenzenecarboximidamide monohydrochloride,benzenecarboximidamide, 4-chloro-, monohydrochloride,4-chlorobenzimidamide hydrochloride,benzamide, p-chloro-, hydrochloride 7ci,benzamidine, p-chloro-, monohydrochloride
203436
4-chlorobenzenecarboximidamide;hydrochloride
C1=CC(=CC=C1C(=N)N)Cl.Cl
14401-51-5
C7H7ClN2·ClH
191.06
RXAOGVQDNBYURA-UHFFFAOYSA-N
4-chlorobenzamidine hydrochloride,4-chlorobenzene-1-carboximidamide hydrochloride,4-chlorobenzamidine hcl,p-chlorobenzamidine hydrochloride,4-chloro-benzamidine hydrochloride,4-chlorobenzenecarboximidamide monohydrochloride,benzenecarboximidamide, 4-chloro-, monohydrochloride,4-chlorobenzimidamide hydrochloride,benzamide, p-chloro-, hydrochloride 7ci,benzamidine, p-chloro-, monohydrochloride
203436
4-chlorobenzenecarboximidamide;hydrochloride
C1=CC(=CC=C1C(=N)N)Cl.Cl
Melting Point 244.0°C to 245.0°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 94 to max. 106% min. (Argentometry)
Linear Formula ClC6H4C(=NH)NH2·HCl
Formula Weight 191.06
Percent Purity 95%
Chemical Name or Material 4-Chlorobenzamidine hydrochloride