Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorobenzamidine hydroiodide, 97%, Acros Organics

Click to view available options
Quantity:
1g
115297-57-9
C7H8ClIN2
282.509
MFCD00051987
YMEVGWRZEYGNJO-UHFFFAOYSA-N
4-chlorobenzamidine hydroiodide,4-chlorobenzimidamide hydroiodide,4-chlorobenzamidine hydriodide,chlorophenamidine hydroiodide,4-chlorobenzenecarboximidamide hydroiodide,acmc-1bu33,benzenecarboximidamide,4-chloro-,hydriodide 1:1,benzenecarboximidamide,4-chloro-, hydriodide 1:1
2734221
4-chlorobenzenecarboximidamide;hydroiodide
C1=CC(=CC=C1C(=N)N)Cl.I
115297-57-9
C7H8ClIN2
282.509
MFCD00051987
YMEVGWRZEYGNJO-UHFFFAOYSA-N
4-chlorobenzamidine hydroiodide,4-chlorobenzimidamide hydroiodide,4-chlorobenzamidine hydriodide,chlorophenamidine hydroiodide,4-chlorobenzenecarboximidamide hydroiodide,acmc-1bu33,benzenecarboximidamide,4-chloro-,hydriodide 1:1,benzenecarboximidamide,4-chloro-, hydriodide 1:1
2734221
4-chlorobenzenecarboximidamide;hydroiodide
C1=CC(=CC=C1C(=N)N)Cl.I
Quantity 1g
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 228°C to 233°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 282.51
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-chlorobenzamidine hydroiodide, 97%
Show More Show Less