Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzenesulfinic acid sodium salt hydrate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
14752-66-0
C6H4ClNaO2S
198.596
MFCD00035602
JFXAUUFCZJYLJF-UHFFFAOYSA-M
sodium 4-chlorobenzenesulfinate,4-chlorobenzenesulfinic acid sodium salt,sodium 4-chlorobenzene sulfinate,unii-x2u129fz4n,sodium p-chlorobenzenesulphinate,sodium 4-chlorobenzene-1-sulfinate,4-chlorobenzene sulfinic acid sodium,4-chlorobenzenesulfinic acid sodium salt hydrate,p-chlorobenzenesulfinic acid, sodium salt,acmc-1cin1
23664783
sodium;4-chlorobenzenesulfinate
C1=CC(=CC=C1S(=O)[O-])Cl.[Na+]
14752-66-0
C6H4ClNaO2S
198.596
MFCD00035602
JFXAUUFCZJYLJF-UHFFFAOYSA-M
sodium 4-chlorobenzenesulfinate,4-chlorobenzenesulfinic acid sodium salt,sodium 4-chlorobenzene sulfinate,unii-x2u129fz4n,sodium p-chlorobenzenesulphinate,sodium 4-chlorobenzene-1-sulfinate,4-chlorobenzene sulfinic acid sodium,4-chlorobenzenesulfinic acid sodium salt hydrate,p-chlorobenzenesulfinic acid, sodium salt,acmc-1cin1
23664783
sodium;4-chlorobenzenesulfinate
C1=CC(=CC=C1S(=O)[O-])Cl.[Na+]
Infrared Spectrum Authentic
Quantity 5g
Color White
Water 14% max. (K.F.)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 198.6
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material 4-Chlorobenzenesulfinic acid sodium salt hydrate
Show More Show Less

EINECSNumber : 238-811-9