Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Chlorobenzhydrol, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Quantity | 100g |
Boiling Point | 150°C to 151°C (2.6mmHg) |
Melting Point | 58°C to 62°C |
Beilstein | 2050235 |
Formula Weight | 218.68 |
Percent Purity | 98% |
Chemical Name or Material | 4-Chlorobenzhydrol |
Safety and Handling
EINECSNumber : 204-333-4
TSCA : Yes
Recommended Storage : Ambient temperatures