Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorobenzhydryl chloride, 98%

Click to view available options
Quantity:
25g
100g
500g
134-83-8
C13H10Cl2
237.123
MFCD00000856
ALKWTKGPKKAZMN-UHFFFAOYSA-N
4-chlorobenzhydryl chloride,4-chlorobenzhydrylchloride,p-chlorobenzhydryl chloride,1-chloro-4-chloro phenyl methyl benzene,benzene, 1-chloro-4-chlorophenylmethyl,chloro 4-chlorophenyl phenylmethane,chloro p-chlorophenyl phenylmethane,methane, chloro p-chlorophenyl phenyl,1-chloro-4-chlorophenylmethyl benzene
241584
1-chloro-4-[chloro(phenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
134-83-8
C13H10Cl2
237.123
MFCD00000856
ALKWTKGPKKAZMN-UHFFFAOYSA-N
4-chlorobenzhydryl chloride,4-chlorobenzhydrylchloride,p-chlorobenzhydryl chloride,1-chloro-4-chloro phenyl methyl benzene,benzene, 1-chloro-4-chlorophenylmethyl,chloro 4-chlorophenyl phenylmethane,chloro p-chlorophenyl phenylmethane,methane, chloro p-chlorophenyl phenyl,1-chloro-4-chlorophenylmethyl benzene
241584
1-chloro-4-[chloro(phenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
Flash Point 194°C (381°F)
Boiling Point 159°C to 160°C (2mmHg)
Quantity 100g
UN Number UN3265
Beilstein 2050236
Formula Weight 237.13
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzhydryl chloride

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA