Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
623-03-0
C7H4ClN
137.57
MFCD00001813
GJNGXPDXRVXSEH-UHFFFAOYSA-N
p-chlorobenzonitrile,benzonitrile, 4-chloro,p-cyanochlorobenzene,benzonitrile, p-chloro,4-cyanochlorobenzene,nitril kyseliny p-chlorbenzoove,p-chlorbenzonitrile,unii-4z0hgp3a8a,pcbn,4-chloro benzonitrile
12163
4-chlorobenzonitrile
C1=CC(=CC=C1C#N)Cl
623-03-0
C7H4ClN
137.57
MFCD00001813
GJNGXPDXRVXSEH-UHFFFAOYSA-N
p-chlorobenzonitrile,benzonitrile, 4-chloro,p-cyanochlorobenzene,benzonitrile, p-chloro,4-cyanochlorobenzene,nitril kyseliny p-chlorbenzoove,p-chlorbenzonitrile,unii-4z0hgp3a8a,pcbn,4-chloro benzonitrile
12163
4-chlorobenzonitrile
C1=CC(=CC=C1C#N)Cl
Boiling Point 223°C
Flash Point 108°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 0.1% max.
Melting Point 91°C to 94°C
Density 108g/mL
Color White to Yellow
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H4CN
Beilstein 09, 341
Solubility Information Solubility in water: insoluble
Formula Weight 137.57
Physical Form Flakes or Chunks
Percent Purity ≥98.5%
Chemical Name or Material 4-Chlorobenzonitrile
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning