Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorobenzotrichloride, 98+%

Click to view available options
Quantity:
25g
100g
500g
5216-25-1
C7H4Cl4
229.909
MFCD00000790
LVZPKYYPPLUECL-UHFFFAOYSA-N
4-chlorobenzotrichloride,1-chloro-4-trichloromethyl benzene,p-chlorobenzotrichloride,benzene, 1-chloro-4-trichloromethyl,p-chlorophenyltrichloromethane,p-trichloromethylchlorobenzene,alpha,alpha,alpha,4-tetrachlorotoluene,unii-1up9gju33s,ccris 8902,p,alpha,alpha,alpha-tetrachlorotoluene
21277
1-chloro-4-(trichloromethyl)benzene
C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl
5216-25-1
C7H4Cl4
229.909
MFCD00000790
LVZPKYYPPLUECL-UHFFFAOYSA-N
4-chlorobenzotrichloride,1-chloro-4-trichloromethyl benzene,p-chlorobenzotrichloride,benzene, 1-chloro-4-trichloromethyl,p-chlorophenyltrichloromethane,p-trichloromethylchlorobenzene,alpha,alpha,alpha,4-tetrachlorotoluene,unii-1up9gju33s,ccris 8902,p,alpha,alpha,alpha-tetrachlorotoluene
21277
1-chloro-4-(trichloromethyl)benzene
C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl
Boiling Point 245°C
Quantity 500g
Flash Point 140°C (284°F)
Melting Point 5°C to 6°C
UN Number UN1760
Beilstein 1866549
Formula Weight 229.92
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzotrichloride

GHS H Statement
H350-H372-H361-H302-H312-H315-H335
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

RTECSNumber : XT8580000

TSCA : TSCA