Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzoyl chloride, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
2.5L
Packaging:
Glass bottle
122-01-0
C7H4Cl2O
175.01
RKIDDEGICSMIJA-UHFFFAOYSA-N
benzoyl chloride, 4-chloro,p-chlorobenzoyl chloride,para-chlorobenzoyl chloride,4-chlorobenzoic acid chloride,4-chlorobenzoylchloride,4-chloro benzoyl chloride,benzoylchloride, 4-chloro,benzoyl chloride, p-chloro,ccris 8935,pcboc
8501
CHEBI:60716
4-chlorobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)Cl
122-01-0
C7H4Cl2O
175.01
RKIDDEGICSMIJA-UHFFFAOYSA-N
benzoyl chloride, 4-chloro,p-chlorobenzoyl chloride,para-chlorobenzoyl chloride,4-chlorobenzoic acid chloride,4-chlorobenzoylchloride,4-chloro benzoyl chloride,benzoylchloride, 4-chloro,benzoyl chloride, p-chloro,ccris 8935,pcboc
8501
CHEBI:60716
4-chlorobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)Cl
Specific Gravity 1.37
Packaging Glass bottle
Refractive Index 1.5770 to 1.5790
Boiling Point 222.0°C
Quantity 2.5L
Infrared Spectrum Authentic
Color Colorless
Melting Point 12.0°C to 14.0°C
Flash Point 105°C
Density 1.3700g/mL
Assay Percent Range 99+%
Linear Formula ClC6H4COCl
Beilstein 09,341
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material 4-Chlorobenzoyl chloride
Show More Show Less