Learn More
4-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals
Description
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.
Solubility
Soluble in water (2.5 mg/ml at 20°C), and methanol.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Melting Point | 68°C to 72°C |
Boiling Point | 234°C |
Quantity | 25 g |
Beilstein | 636502 |
Solubility Information | Soluble in water (2.5mg/ml at 20°C),and methanol. |
Formula Weight | 142.59 |
Percent Purity | 99% |
Chemical Name or Material | 4-Chlorobenzyl alcohol |
Safety and Handling
EINECSNumber | 212-852-2 |
RTECSNumber | DN9242000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.