Alfa Aesar™ 4-Chlorobenzyl alcohol, 99%

Manufacturer: Alfa Aesar™ A1150436

Quantity 500g

 View more versions of this product

Catalog No. AAA1150436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 873-76-7
    Molecular Formula C7H7ClO
    Formula Weight 142.59
    MDL Number MFCD00004652
    Beilstein 636502
    Chemical Name or Material 4-Chlorobenzyl alcohol

    Specifications

    Formula Weight 142.59
    CAS 873-76-7
    Quantity 500g
    Molecular Formula C7H7ClO
    MDL Number MFCD00004652
    Beilstein 636502
    RTECS Number DN9242000
    Boiling Point 234°C
    Melting Point 68° to 72°C
    Chemical Name or Material 4-Chlorobenzyl alcohol
    Assay 99%

    Description

    Applications
    4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.

    Solubility
    Soluble in water (2.5 mg/ml at 20°C), and methanol.

    Notes
    Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit