Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorobenzyl bromide, 98+%

Click to view available options
Quantity:
5g
25g
100g
622-95-7
C7H6BrCl
205.479
MFCD00040714
KQNBRMUBPRGXSL-UHFFFAOYSA-N
4-chlorobenzyl bromide,1-bromomethyl-4-chlorobenzene,p-chlorobenzyl bromide,4-chlorobenzylbromide,benzene, 1-bromomethyl-4-chloro,alpha-bromo-4-chlorotoluene,4cbb,p-chlorobenzylbromide,4-chloro-benzylbromide
69329
1-(bromomethyl)-4-chlorobenzene
C1=CC(=CC=C1CBr)Cl
622-95-7
C7H6BrCl
205.479
MFCD00040714
KQNBRMUBPRGXSL-UHFFFAOYSA-N
4-chlorobenzyl bromide,1-bromomethyl-4-chlorobenzene,p-chlorobenzyl bromide,4-chlorobenzylbromide,benzene, 1-bromomethyl-4-chloro,alpha-bromo-4-chlorotoluene,4cbb,p-chlorobenzylbromide,4-chloro-benzylbromide
69329
1-(bromomethyl)-4-chlorobenzene
C1=CC(=CC=C1CBr)Cl
Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
Quantity 5g
Melting Point 46°C to 51°C
Odor Pungent
Boiling Point 107°C to 108°C (8mmHg)
UN Number UN3261
Beilstein 606497
Formula Weight 205.49
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.