Alfa Aesar™ 4-Chlorobenzyl bromide, 98+%

Manufacturer: Alfa Aesar™ B2285814

Quantity 25g

 View more versions of this product

Catalog No. AAB2285814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 622-95-7
    Molecular Formula C7H6BrCl
    Formula Weight 205.49
    MDL Number MFCD00040714
    UN Number UN3261
    Beilstein 606497
    Synonym alpha-Bromo-4-chlorotoluene
    Chemical Name or Material 4-Chlorobenzyl bromide

    Specifications

    Formula Weight 205.49
    Odor Pungent
    UN Number UN3261
    Beilstein 606497
    Synonym alpha-Bromo-4-chlorotoluene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point 46° to 51°C
    Quantity 25g
    Molecular Formula C7H6BrCl
    MDL Number MFCD00040714
    CAS 622-95-7
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 107° to 108°C (8mmHg)
    Chemical Name or Material 4-Chlorobenzyl bromide
    Assay ≥98%

    Description

    Applications
    4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit