Alfa Aesar™ 4-Chlorobenzylmagnesium chloride, 0.50M in 2-Me-THF

Manufacturer: Alfa Aesar™ H54625AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54625AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 874-72-6
    Molecular Formula C7H6Cl2Mg
    Formula Weight 185.33
    MDL Number MFCD01311474
    UN Number UN2924
    Chemical Name or Material 4-Chlorobenzylmagnesium chloride

    Specifications

    Formula Weight 185.33
    Molecular Formula C7H6Cl2Mg
    MDL Number MFCD01311474
    CAS 874-72-6
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 0.50M in 2-MeTHF
    UN Number UN2924
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Quantity 100mL
    Chemical Name or Material 4-Chlorobenzylmagnesium chloride

    Description

    Applications
    4-Chlorobenzylmagnesium chloride is used as an intermediate in organic synthesis. It is used as a precursor for the preparation of 1-chloro-4-(4-methyl-3-pentenyl)benzene by reacting with 1-bromo-3-methyl-2-butene.

    Solubility
    Miscible with tetrahydrofuran.

    Notes
    Air and moisture sensitive. Incompatible with acids, acid chlorides, alcohols, oxidizing agents and chloroformates.

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit