Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chlorobiphenyl-4-sulfonyl chloride, 97%, Acros Organics™

Click to view available options
Quantity:
1g
10g
20443-74-7
C12H8Cl2O2S
287.17
NWYUSJMIHFIMTA-UHFFFAOYSA-N
4'-chlorobiphenyl-4-sulfonyl chloride,4-4-chlorophenyl benzenesulfonyl chloride,4'-chloro-biphenyl-4-sulfonyl chloride,4'-chloro-1,1'-biphenyl-4-sulfonyl chloride,4'-chloro-4-biphenylylsulphonyl chloride,4'-chloro 1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 4'-chloro,4'-chlorobiphenyl4-sulfonyl chloride,4-chlorobiphenyl-4'-sulfonyl chloride,4'-chloro biphenyl-4-sulfonyl chloride
2794745
4-(4-chlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)Cl
20443-74-7
C12H8Cl2O2S
287.17
NWYUSJMIHFIMTA-UHFFFAOYSA-N
4'-chlorobiphenyl-4-sulfonyl chloride,4-4-chlorophenyl benzenesulfonyl chloride,4'-chloro-biphenyl-4-sulfonyl chloride,4'-chloro-1,1'-biphenyl-4-sulfonyl chloride,4'-chloro-4-biphenylylsulphonyl chloride,4'-chloro 1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 4'-chloro,4'-chlorobiphenyl4-sulfonyl chloride,4-chlorobiphenyl-4'-sulfonyl chloride,4'-chloro biphenyl-4-sulfonyl chloride
2794745
4-(4-chlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)Cl
Quantity 10g
Melting Point 109.0°C to 110.0°C
Assay Percent Range 96% min. as diethylsulfonamide (HPLC)
Formula Weight 287.17
Percent Purity 97%
Chemical Name or Material 4'-Chlorobiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger