Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorobutyl acetate, 98%

Click to view available options
Quantity:
25g
100g
500g
6962-92-1
C6H11ClO2
150.602
MFCD00001013
PYLDCZJUHYVOAF-UHFFFAOYSA-N
1-acetoxy-4-chlorobutane,4-chloro-n-butyl acetate,1-chloro-4-acetoxybutane,1-butanol, 4-chloro-, acetate,4-chlorobutylacetate,4-chloro-1-butanol acetate,4-chlorbut-1-ylacetat,delta-chlorobutyl acetate,1-butanol, 4-chloro-, 1-acetate,4-chlorbut-1-ylacetat german
23399
4-chlorobutyl acetate
CC(=O)OCCCCCl
6962-92-1
C6H11ClO2
150.602
MFCD00001013
PYLDCZJUHYVOAF-UHFFFAOYSA-N
1-acetoxy-4-chlorobutane,4-chloro-n-butyl acetate,1-chloro-4-acetoxybutane,1-butanol, 4-chloro-, acetate,4-chlorobutylacetate,4-chloro-1-butanol acetate,4-chlorbut-1-ylacetat,delta-chlorobutyl acetate,1-butanol, 4-chloro-, 1-acetate,4-chlorbut-1-ylacetat german
23399
4-chlorobutyl acetate
CC(=O)OCCCCCl
Flash Point 64°C (147°F)
Boiling Point 81°C to 83°C (11mmHg)
Quantity 500g
Beilstein 1749681
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material 4-Chlorobutyl acetate

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EL1328000

TSCA : TSCA