Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorobutyronitrile, 97%

Click to view available options
Quantity:
50g
250g
1000g
628-20-6
C4H6ClN
103.549
MFCD00001972
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutyronitrile,butanenitrile, 4-chloro,butyronitrile, 4-chloro,gamma-chlorobutyronitrile,1-chloro-3-cyanopropane,4-chloro-butyronitrile,.gamma.-chlorobutyronitrile,4chlorobutyronitrile,4-chlorobutyroonitrile,4-chloro-butanenitrile
12336
4-chlorobutanenitrile
C(CC#N)CCl
628-20-6
C4H6ClN
103.549
MFCD00001972
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutyronitrile,butanenitrile, 4-chloro,butyronitrile, 4-chloro,gamma-chlorobutyronitrile,1-chloro-3-cyanopropane,4-chloro-butyronitrile,.gamma.-chlorobutyronitrile,4chlorobutyronitrile,4-chlorobutyroonitrile,4-chloro-butanenitrile
12336
4-chlorobutanenitrile
C(CC#N)CCl
Refractive Index 1.444
Density 1.084
Flash Point 85°C (185°F)
Boiling Point 195°C to 197°C
Quantity 50g
Linear Formula Cl(CH2)3CN
UN Number UN3276
Beilstein 1633582
Formula Weight 103.55
Percent Purity 97%
Chemical Name or Material 4-Chlorobutyronitrile
Show More Show Less

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 211-031-6

RTECSNumber : ET9020000

TSCA : Yes

Recommended Storage : Ambient temperatures