Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorochalcone, 98+%

Click to view available options
Quantity:
10g
50g
250g
956-04-7
C15H11ClO
242.702
MFCD00016345
ABGIIXRNMHUKII-DHZHZOJOSA-N
4-chlorochalcone,p-chlorochalcone,p-chlorostyryl phenyl ketone,3-4-chlorophenyl-1-phenyl-2-propen-1-one,4-chlorostyryl phenyl ketone,3-4-chlorophenyl-1-phenylprop-2-en-1-one,2e-3-4-chlorophenyl-1-phenylprop-2-en-1-one,4-chlorobenzylidene acetophenone,e-3-4-chlorophenyl-1-phenylprop-2-en-1-one,chalcone, 4-chloro-6ci,7ci,8ci
5377022
CHEBI:34398
(E)-3-(4-chlorophenyl)-1-phenylprop-2-en-1-one
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)Cl
956-04-7
C15H11ClO
242.702
MFCD00016345
ABGIIXRNMHUKII-DHZHZOJOSA-N
4-chlorochalcone,p-chlorochalcone,p-chlorostyryl phenyl ketone,3-4-chlorophenyl-1-phenyl-2-propen-1-one,4-chlorostyryl phenyl ketone,3-4-chlorophenyl-1-phenylprop-2-en-1-one,2e-3-4-chlorophenyl-1-phenylprop-2-en-1-one,4-chlorobenzylidene acetophenone,e-3-4-chlorophenyl-1-phenylprop-2-en-1-one,chalcone, 4-chloro-6ci,7ci,8ci
5377022
CHEBI:34398
(E)-3-(4-chlorophenyl)-1-phenylprop-2-en-1-one
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)Cl
Quantity 250g
Melting Point 114°C to 116°C
Beilstein 1105953
Formula Weight 242.71
Percent Purity ≥98%
Chemical Name or Material 4-Chlorochalcone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-476-1

RTECSNumber : UD5572994

TSCA : No

Recommended Storage : Ambient temperatures