Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloroindole, 97%

Click to view available options
Quantity:
1g
5g
25235-85-2
C8H6ClN
151.593
MFCD00005665
SVLZRCRXNHITBY-UHFFFAOYSA-N
4-chloroindole,1h-indole, 4-chloro,4-chloro indole,1h-indole, 4-chloro-,conjugate acid 1:1,4-chloro-indole,zlchem 277,indole, 4-chloro,pubchem7232,4-chloranyl-1h-indole,acmc-209gia
91345
4-chloro-1H-indole
C1=CC2=C(C=CN2)C(=C1)Cl
25235-85-2
C8H6ClN
151.593
MFCD00005665
SVLZRCRXNHITBY-UHFFFAOYSA-N
4-chloroindole,1h-indole, 4-chloro,4-chloro indole,1h-indole, 4-chloro-,conjugate acid 1:1,4-chloro-indole,zlchem 277,indole, 4-chloro,pubchem7232,4-chloranyl-1h-indole,acmc-209gia
91345
4-chloro-1H-indole
C1=CC2=C(C=CN2)C(=C1)Cl
Quantity 1g
Refractive Index 1.6280
Odor Odorless
Boiling Point 129°C to 130°C (4mmHg)
Density 1.259g/mL
Beilstein 114880
Formula Weight 151.59
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Chloroindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning