Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroindole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
25235-85-2
C8H6ClN
151.593
SVLZRCRXNHITBY-UHFFFAOYSA-N
4-chloroindole,1h-indole, 4-chloro,4-chloro indole,1h-indole, 4-chloro-,conjugate acid 1:1,4-chloro-indole,zlchem 277,indole, 4-chloro,pubchem7232,4-chloranyl-1h-indole,acmc-209gia
91345
4-chloro-1H-indole
C1=CC2=C(C=CN2)C(=C1)Cl
25235-85-2
C8H6ClN
151.593
SVLZRCRXNHITBY-UHFFFAOYSA-N
4-chloroindole,1h-indole, 4-chloro,4-chloro indole,1h-indole, 4-chloro-,conjugate acid 1:1,4-chloro-indole,zlchem 277,indole, 4-chloro,pubchem7232,4-chloranyl-1h-indole,acmc-209gia
91345
4-chloro-1H-indole
C1=CC2=C(C=CN2)C(=C1)Cl
Color Yellow
Packaging Glass bottle
Quantity 1g
Density 1.2500g/mL
Infrared Spectrum Authentic
Refractive Index 1.6270 to 1.6290 (20°C, 589nm)
Assay Percent Range 99%
Formula Weight 151.6
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4-Chloroindole
Show More Show Less

Recommended Storage : May darken on storage