Alfa Aesar™ 4-(Chloromethyl)-1,2-diphenylethane, 98%

Manufacturer: Alfa Aesar™ B2522203

Quantity 1g

 View more versions of this product

Catalog No. AAB2522203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80676-35-3
    Molecular Formula C15H15Cl
    Formula Weight 230.73
    MDL Number MFCD02093918
    UN Number UN1759
    Synonym 4-(Chloromethyl)bibenzyl; 4-(Chloromethyl)dibenzyl
    Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane

    Specifications

    Formula Weight 230.73
    Molecular Formula C15H15Cl
    MDL Number MFCD02093918
    CAS 80676-35-3
    UN Number UN1759
    Synonym 4-(Chloromethyl)bibenzyl; 4-(Chloromethyl)dibenzyl
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Melting Point 38° to 42°C
    Quantity 1g
    Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit