Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-(Chloromethyl)benzonitrile, 98+%
Click to view available options
Quantity:
10g
50g
250g
Specifications
Quantity | 10g |
Melting Point | 77°C to 80°C |
UN Number | UN2923 |
Formula Weight | 151.6 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-(Chloromethyl)benzonitrile |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c
H301+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-869-5
TSCA : No
Recommended Storage : Ambient temperatures