Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloronicotinic acid, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
10177-29-4
C6H4ClNO2
157.56
IMRGVWZLCZERSQ-UHFFFAOYSA-N
4-chloronicotinic acid,4-chloronicotinicacid,4-chloro-3-pyridinecarboxylic acid,4-chloro-nicotinic acid,3-pyridinecarboxylic acid, 4-chloro,3-carboxy-4-chloropyridine,4-chlornicotinsaure,pubchem12989,acmc-1br3v,4-chloropyridine-3-carboxyl
818229
4-chloropyridine-3-carboxylic acid
C1=CN=CC(=C1Cl)C(=O)O
10177-29-4
C6H4ClNO2
157.56
IMRGVWZLCZERSQ-UHFFFAOYSA-N
4-chloronicotinic acid,4-chloronicotinicacid,4-chloro-3-pyridinecarboxylic acid,4-chloro-nicotinic acid,3-pyridinecarboxylic acid, 4-chloro,3-carboxy-4-chloropyridine,4-chlornicotinsaure,pubchem12989,acmc-1br3v,4-chloropyridine-3-carboxyl
818229
4-chloropyridine-3-carboxylic acid
C1=CN=CC(=C1Cl)C(=O)O
Melting Point 170°C to 171°C
Quantity 5g
Assay Percent Range 95% min. (HPLC)
Formula Weight 157.56
Percent Purity 96%
Chemical Name or Material 4-Chloronicotinic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning