Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorophenoxyacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
3598-13-8
C8H6ClNO
167.592
YUGDKEWUYZXXRU-UHFFFAOYSA-N
2-4-chlorophenoxy acetonitrile,4-chlorophenoxyacetonitrile,4-chlorophenoxy acetonitrile,4-chloro-phenoxy-acetonitrile,p-chlorophenoxyacetonitrile,1-chloro-4-cyanomethoxy benzene,acetonitrile, 4-chlorophenoxy,4-chloro-o-cyanomethylphenol,2-4-chlorophenoxy ethanenitrile,acmc-20aopp
306444
2-(4-chlorophenoxy)acetonitrile
C1=CC(=CC=C1OCC#N)Cl
3598-13-8
C8H6ClNO
167.592
YUGDKEWUYZXXRU-UHFFFAOYSA-N
2-4-chlorophenoxy acetonitrile,4-chlorophenoxyacetonitrile,4-chlorophenoxy acetonitrile,4-chloro-phenoxy-acetonitrile,p-chlorophenoxyacetonitrile,1-chloro-4-cyanomethoxy benzene,acetonitrile, 4-chlorophenoxy,4-chloro-o-cyanomethylphenol,2-4-chlorophenoxy ethanenitrile,acmc-20aopp
306444
2-(4-chlorophenoxy)acetonitrile
C1=CC(=CC=C1OCC#N)Cl
Melting Point 45°C to 49°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Tan-Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4OCH2CN
Formula Weight 167.6
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-chlorophenoxyacetonitrile
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning