Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chlorophenoxyacetyl Chloride 98.0+%, TCI America™
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Color | Yellow |
Boiling Point | 165°C |
UN Number | 3265 |
Quantity | 100 g |
Formula Weight | 205.03 |
Percent Purity | ≥98.0% (T) |
Physical Form | Liquid |
Chemical Name or Material | 4-Chlorophenoxyacetyl Chloride |
Safety and Handling
TSCA : No