Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorophenyl chloroformate, 95%, Thermo Scientific™

Catalog No. AC435660050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-45-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
InChI Key RYWGPCLTVXMMHO-UHFFFAOYSA-N
Synonym 4-chlorophenyl chloroformate, 4-chlorophenylchloroformate, p-chlorophenyl chloroformate, carbonochloridic acid, 4-chlorophenyl ester, 4-chlorophenyl carbonochloridate, chloroformic acid 4-chlorophenyl ester, acmc-1biw7, 4-chloro-phenyl chloroformate, ksc493g4j, 4-chlorophenylcarbonochloridate
PubChem CID 139059
IUPAC Name (4-chlorophenyl) carbonochloridate
SMILES C1=CC(=CC=C1OC(=O)Cl)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.3650g/mL
Boiling Point 100°C to 102°C (16.0 hPa)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.365
Formula Weight 191.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-chlorophenyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.