Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorophenyl cyclopropyl ketone, 98%

Click to view available options
Quantity:
10g
50g
250g
6640-25-1
C10H9ClO
180.631
MFCD00001295
OPSFCTBBDIDFJM-UHFFFAOYSA-N
4-chlorophenyl cyclopropyl ketone,4-chlorophenyl cyclopropyl methanone,4-chlorobenzoyl cyclopropane,4-chlorobenzoylcyclopropane,methanone, 4-chlorophenyl cyclopropyl,4-chlorophenylcyclopropylmethanone,pubchem16774,acmc-209nvc,cyclopropyl 4-chlorophenyl ketone
81148
(4-chlorophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC=C(C=C2)Cl
6640-25-1
C10H9ClO
180.631
MFCD00001295
OPSFCTBBDIDFJM-UHFFFAOYSA-N
4-chlorophenyl cyclopropyl ketone,4-chlorophenyl cyclopropyl methanone,4-chlorobenzoyl cyclopropane,4-chlorobenzoylcyclopropane,methanone, 4-chlorophenyl cyclopropyl,4-chlorophenylcyclopropylmethanone,pubchem16774,acmc-209nvc,cyclopropyl 4-chlorophenyl ketone
81148
(4-chlorophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC=C(C=C2)Cl
Melting Point 29°C to 31°C
Flash Point >110°C (230°F)
Quantity 10g
Boiling Point 135°C to 137°C (10mmHg)
Beilstein 2044845
Formula Weight 180.63
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl cyclopropyl ketone