Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorophenyl cyclopropyl ketone, 98%

Manufacturer:  Alfa Aesar™ B2276618

Quantity 50g

 View more versions of this product

Catalog No. AAB2276618


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6640-25-1
180.63
2044845
4-Chlorophenyl cyclopropyl ketone
C10H9ClO
MFCD00001295
(4-Chlorobenzoyl)cyclopropane

Specifications

>110°C (230°F)
29° to 31°C
6640-25-1
180.63
2044845
4-Chlorophenyl cyclopropyl ketone
135° to 137°C (10mmHg)
50g
C10H9ClO
MFCD00001295
(4-Chlorobenzoyl)cyclopropane
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit