Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl dichlorophosphate, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
772-79-2
C6H4Cl3O2P
245.43
MFCD00009705
CCZMQYGSXWZFKI-UHFFFAOYSA-N
4-chlorophenyl phosphorodichloridate,p-chlorophenyl dichlorophosphate,4-chlorophenyl dichlorophosphate,4-chlorophenylphosphorodichloridate,phosphorodichloridic acid, 4-chlorophenyl ester,4-chlorophenylphosphoryl dichloride,acmc-1bjgo,cczmqygsxwzfki-uhfffaoysa,4-chlorophenyl dichlorophosphinate,4-chlorophenyl dichloridophosphate #
69879
1-chloro-4-dichlorophosphoryloxybenzene
C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl
772-79-2
C6H4Cl3O2P
245.43
MFCD00009705
CCZMQYGSXWZFKI-UHFFFAOYSA-N
4-chlorophenyl phosphorodichloridate,p-chlorophenyl dichlorophosphate,4-chlorophenyl dichlorophosphate,4-chlorophenylphosphorodichloridate,phosphorodichloridic acid, 4-chlorophenyl ester,4-chlorophenylphosphoryl dichloride,acmc-1bjgo,cczmqygsxwzfki-uhfffaoysa,4-chlorophenyl dichlorophosphinate,4-chlorophenyl dichloridophosphate #
69879
1-chloro-4-dichlorophosphoryloxybenzene
C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl
Specific Gravity 1.508
Packaging Glass bottle
Flash Point 106°C
Refractive Index 1.5380 to 1.5400
Infrared Spectrum Authentic
Color Colorless
Boiling Point 114.0°C to 116.0°C (1.0 mmHg)
Density 1.5080g/mL
Quantity 5g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula Cl2P(O)OC6H4Cl
Formula Weight 245.43
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenyl dichlorophosphate, 98+%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger