Alfa Aesar™ (4-Chlorophenylethynyl)trimethylsilane, 97%

Manufacturer: Alfa Aesar™ H5512914

Quantity 25g

 View more versions of this product

Catalog No. AAH5512914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78704-49-1
    Formula Weight 208.76
    MDL Number MFCD03427267
    Chemical Name or Material (4-Chlorophenylethynyl)trimethylsilane

    Specifications

    Formula Weight 208.76
    Melting Point 47° to 51°C
    Physical Form Solid
    MDL Number MFCD03427267
    Color White to pale brown
    Quantity 25g
    CAS 78704-49-1
    Chemical Name or Material (4-Chlorophenylethynyl)trimethylsilane
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit