Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
873-77-8,109-99-9,108-88-3
C6H4BrClMg
215.757
MFCD00009926
DLJIPJMUYCUTOV-UHFFFAOYSA-M
4-chlorophenylmagnesium bromide,bromo p-chlorophenyl magnesium,bromo 4-chlorophenyl magnesium,p-chlorophenylmagnesium bromide,4-chlorophenyl magnesium bromide,pubchem24069,4-chlorophenylmagnesiumbromide,4-chlorphenylmagnesium bromide,4-chloro phenylmagnesiumbromide,4-chlorophenyhnagnesium bromide
101888
magnesium;chlorobenzene;bromide
C1=CC(=CC=[C-]1)Cl.[Mg+2].[Br-]
873-77-8,109-99-9,108-88-3
C6H4BrClMg
215.757
MFCD00009926
DLJIPJMUYCUTOV-UHFFFAOYSA-M
4-chlorophenylmagnesium bromide,bromo p-chlorophenyl magnesium,bromo 4-chlorophenyl magnesium,p-chlorophenylmagnesium bromide,4-chlorophenyl magnesium bromide,pubchem24069,4-chlorophenylmagnesiumbromide,4-chlorphenylmagnesium bromide,4-chloro phenylmagnesiumbromide,4-chlorophenyhnagnesium bromide
101888
magnesium;chlorobenzene;bromide
C1=CC(=CC=[C-]1)Cl.[Mg+2].[Br-]
CAS Min % 21.58,76.27
CAS Max % 23.73,78.42
Appearance of Solution Yellowish-green solution
Linear Formula Cl(C6H6)MgBr
Concentration 1M, exact strength on the certificate of analysis
Density 1.0000g/mL
Formula Weight 215.76
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 1
Quantity 800mL
Flash Point −21°C
Chemical Name or Material 4-Chlorophenylmagnesium bromide
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of damaging the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger