Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylsulfonylacetone, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
5000-48-6
C9H9ClO3S
232.69
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone,1-4-chlorophenyl sulfonyl acetone,1-4-chlorophenyl sulfonyl propan-2-one,4-chlorobenzenesulphonylacetone,p-chlorophenylsulfonylacetone,1-4-chlorobenzenesulfonyl propan-2-one,4-chlorophenylsulfonyl acetone,1-4-chlorophenyl sulfonylpropan-2-one,maybridge1_004685
521269
1-(4-chlorophenyl)sulfonylpropan-2-one
CC(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
5000-48-6
C9H9ClO3S
232.69
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone,1-4-chlorophenyl sulfonyl acetone,1-4-chlorophenyl sulfonyl propan-2-one,4-chlorobenzenesulphonylacetone,p-chlorophenylsulfonylacetone,1-4-chlorobenzenesulfonyl propan-2-one,4-chlorophenylsulfonyl acetone,1-4-chlorophenyl sulfonylpropan-2-one,maybridge1_004685
521269
1-(4-chlorophenyl)sulfonylpropan-2-one
CC(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
Packaging Glass Bottle
Melting Point 89°C to 91°C
Quantity 5g
Assay Percent Range 98% min. (GC)
Linear Formula CCl6H4SO2CH2COCH3
Solubility Information Solubility in water: insoluble
Formula Weight 232.69
Percent Purity 98+%
Chemical Name or Material 4-Chlorophenylsulfonylacetone

GHS Signal Word: Warning