Alfa Aesar™ (4-Chlorophenylthio)acetonitrile, 98%

Manufacturer: Alfa Aesar™ A1231914

Quantity 25g

 View more versions of this product

Catalog No. AAA1231914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18527-19-0
    Molecular Formula C8H6ClNS
    Formula Weight 183.66
    MDL Number MFCD00019832
    Chemical Name or Material (4-Chlorophenylthio)acetonitrile

    Specifications

    Formula Weight 183.66
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H6ClNS
    MDL Number MFCD00019832
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    CAS 18527-19-0
    Health Hazard 1 Warning
    Melting Point 79° to 81°C
    Quantity 25g
    Chemical Name or Material (4-Chlorophenylthio)acetonitrile
    Assay 98%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit