Alfa Aesar™ 4-Chlorophenylurea, 98%

Manufacturer: Alfa Aesar™ B2509022

Quantity 100g

 View more versions of this product

Catalog No. AAB2509022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 140-38-5
    Molecular Formula C7H7ClN2O
    Formula Weight 170.6
    MDL Number MFCD00014788
    Beilstein 908492
    Chemical Name or Material 4-Chlorophenylurea

    Specifications

    Formula Weight 170.6
    MDL Number MFCD00014788
    Beilstein 908492
    RTECS Number YS6127000
    Quantity 100g
    Molecular Formula C7H7ClN2O
    CAS 140-38-5
    Melting Point 204° to 205°C
    Chemical Name or Material 4-Chlorophenylurea
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit