Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chlorophenylurea, 98%

Click to view available options
Quantity:
5g
25g
100g
140-38-5
C7H7ClN2O
170.596
MFCD00014788
RECCURWJDVZHIH-UHFFFAOYSA-N
1-4-chlorophenyl urea,4-chlorophenyl urea,urea, 4-chlorophenyl,p-chlorophenyl urea,p-cpu,urea, p-chlorophenyl,1-p-chlorophenyl urea,p-chlorophenylurea,urea, 1-p-chlorophenyl,n-4-chlorophenyl urea
8796
(4-chlorophenyl)urea
C1=CC(=CC=C1NC(=O)N)Cl
140-38-5
C7H7ClN2O
170.596
MFCD00014788
RECCURWJDVZHIH-UHFFFAOYSA-N
1-4-chlorophenyl urea,4-chlorophenyl urea,urea, 4-chlorophenyl,p-chlorophenyl urea,p-cpu,urea, p-chlorophenyl,1-p-chlorophenyl urea,p-chlorophenylurea,urea, 1-p-chlorophenyl,n-4-chlorophenyl urea
8796
(4-chlorophenyl)urea
C1=CC(=CC=C1NC(=O)N)Cl
Melting Point 204°C to 205°C
Quantity 5g
Beilstein 908492
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylurea

RTECSNumber : YS6127000