Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropyrazole, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
15878-00-9
C3H3ClN2
102.52
BADSZRMNXWLUKO-UHFFFAOYSA-N
4-chloropyrazole,1h-pyrazole, 4-chloro,4-chloro pyrazole,1h-pyrazole, 4-chloro-9ci,pyrazole, 4-chloro,zlchem 159,pubchem14005,5-23-04-00164 beilstein handbook reference
27524
4-chloro-1H-pyrazole
C1=C(C=NN1)Cl
15878-00-9
C3H3ClN2
102.52
BADSZRMNXWLUKO-UHFFFAOYSA-N
4-chloropyrazole,1h-pyrazole, 4-chloro,4-chloro pyrazole,1h-pyrazole, 4-chloro-9ci,pyrazole, 4-chloro,zlchem 159,pubchem14005,5-23-04-00164 beilstein handbook reference
27524
4-chloro-1H-pyrazole
C1=C(C=NN1)Cl
Infrared Spectrum Authentic
Melting Point 71.0°C to 73.0°C
Quantity 25g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 102.52
Percent Purity 98%
Chemical Name or Material 4-Chloropyrazole

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning