Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropyridine-N-oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1121-76-2
C5H4ClNO
129.543
MFCD00047425
DPJVRASYWYOFSJ-UHFFFAOYSA-N
4-chloropyridine n-oxide,4-chloropyridine 1-oxide,4-chloropyridine oxide,pyridine, 4-chloro-, 1-oxide,4-chloropyridine-n-oxide,4-chloro-pyridine 1-oxide,4-chloropuridine n-oxide,unii-f3196ueb9q,4-chloropyridin-1-ol,pubchem19817
70724
4-chloro-1-oxidopyridin-1-ium
C1=C[N+](=CC=C1Cl)[O-]
1121-76-2
C5H4ClNO
129.543
MFCD00047425
DPJVRASYWYOFSJ-UHFFFAOYSA-N
4-chloropyridine n-oxide,4-chloropyridine 1-oxide,4-chloropyridine oxide,pyridine, 4-chloro-, 1-oxide,4-chloropyridine-n-oxide,4-chloro-pyridine 1-oxide,4-chloropuridine n-oxide,unii-f3196ueb9q,4-chloropyridin-1-ol,pubchem19817
70724
4-chloro-1-oxidopyridin-1-ium
C1=C[N+](=CC=C1Cl)[O-]
Color White to Yellow
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, IV, 2500
Formula Weight 129.55
Physical Form Powder and Chunks
Percent Purity ≥97.5%
Chemical Name or Material 4-Chloropyridine-N-oxide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.