Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chloropyridine N-Oxide 97.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Quantity | 25 g |
Formula Weight | 129.54 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 4-Chloropyridine N-Oxide |
Safety and Handling
TSCA : No