Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorosalicylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
5106-98-9
C7H5ClO3
172.57
MFCD00002449
LWXFCZXRFBUOOR-UHFFFAOYSA-N
4-chlorosalicylic acid,4-chloro salicylic acid,benzoic acid, 4-chloro-2-hydroxy,2-hydroxy-4-chlorobenzoic acid,4-chloro-2-hydroxy benzoic acid,4-chloro-2-hydroxy-benzoic acid,salicylic acid, 4-chloro,4-chlorosalicylicacid,pubchem3845,4-chloro-salicylic acid
78782
4-chloro-2-hydroxybenzoic acid
C1=CC(=C(C=C1Cl)O)C(=O)O
5106-98-9
C7H5ClO3
172.57
MFCD00002449
LWXFCZXRFBUOOR-UHFFFAOYSA-N
4-chlorosalicylic acid,4-chloro salicylic acid,benzoic acid, 4-chloro-2-hydroxy,2-hydroxy-4-chlorobenzoic acid,4-chloro-2-hydroxy benzoic acid,4-chloro-2-hydroxy-benzoic acid,salicylic acid, 4-chloro,4-chlorosalicylicacid,pubchem3845,4-chloro-salicylic acid
78782
4-chloro-2-hydroxybenzoic acid
C1=CC(=C(C=C1Cl)O)C(=O)O
Melting Point 209°C to 213°C
Color White to Beige
Packaging Plastic bottle
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(OH)CO2H
Beilstein 10, 101
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 172.57
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-chlorosalicylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-829-7